Key Documents
Z376221
Sigma-Aldrich
PCR Workstation
W × D × H 48 in. × 24 in. × 24 in., AC/DC input 240 V AC, Euro plug
About This Item
UNSPSC Code:
41106300
NACRES:
NB.22
Product Z376221 is not currently sold in your country. Contact Technical Service
technique(s)
PCR: suitable
W × D × H
48 in. × 24 in. × 24 in.
General description
A self-contained work area that will help protect your PCR runs against contamination. Dual UV bulbs irradiate the work area prior to use, reducing the possibility that contaminating DNA will be amplified. Containment features reduce the chance of airborne contamination.
The non-ventilated hood has acrylic windows for a clear view, an acid-resistant black formica work surface, a laminated safety glass fascia, and double access doors. The germicidal lamps are controlled by a 24-hr timer. The work light is provided by dual fluorescent lamps, and there is a ceiling-mounted double AC outlet. Replacement part sold separately.
Legal Information
PCR Workstation is a trademark of C.B.S. Scientific Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)