Key Documents
Z337293
BRAND® conical joint stopper, PP
joint: ST/NS 29/32
Select a Size
10 EA
$37.60
$37.60
Estimated to ship on16 April 2025
Select a Size
Change View
10 EA
$37.60
About This Item
UNSPSC Code:
41122500
NACRES:
NB.35
$37.60
Estimated to ship on16 April 2025
Joint
joint: ST/NS29/32
feature
autoclavable
manufacturer/tradename
BRAND
General description
Hollow, with sealing ribs and square grip.
Other Notes
BR144435 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)