Key Documents
Z332399
Bottle adapters for BRAND® Dispensers and Burettes
GL 45 thread, GL 38 bottle, ETFE
Select a Size
1 EA
$51.20
$51.20
Available to ship on15 April 2025Details
Select a Size
Change View
1 EA
$51.20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
$51.20
Available to ship on15 April 2025Details
material
ETFE
manufacturer/tradename
BRAND 704399
fitting
GL 38 bottle
GL 45 thread
Related Categories
General description
For all Dispensette® models as well as seripettor® and QuikSip
Other Notes
BR704399 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)