Z328774
BRAND® magnetic stirring bar, PTFE, cylindrical
L 25 mm, bar diam. 6 mm
Synonym(s):
PTFE Magnetic Stirrer
Select a Size
$73.20
Available to ship on17 April 2025Details
Select a Size
About This Item
$73.20
Available to ship on17 April 2025Details
Recommended Products
material
PTFE-coated
cylindrical
manufacturer/tradename
BRAND 137120
L
25 mm
bar diam.
6 mm
Related Categories
General description
Features and Benefits
- BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
- The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
- Ideal for large volumes or highly viscous mixtures.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)