Z325945
BRAND® BLAUBRAND® ETERNA graduated pipette, calibrated to deliver
type 3, (zero on top), capacity 20 mL , with 0.1 subdivisions, suitable for cotton plugs
Select a Size
$142.00
Estimated to ship on29 May 2025
Select a Size
About This Item
$142.00
Estimated to ship on29 May 2025
Recommended Products
material
AR-Glas®
Agency
according to DIN 12 697
feature
class AS with batch number
graduation
subdivision 0.1 mL
with large suction tube suitable for cotton plugs
packaging
pack of 6
manufacturer/tradename
BRAND 28414
measuring range
accuracy: 0.1 mL
capacity
20 mL , with 0.1 subdivisions
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Accurate volumetric measurement instrument for precision analyses.
- BRAND® BLAUBRAND® ETERNA graduated pipette of capacity 20 mL, type 3 (zero on top), made of AR-GLAS®, is manufactured according to DIN EN ISO 835.
- Calibrated to deliver (TD, Ex).
- Reduced waiting time (5 sec) and simplified handling.
- High volume precision.
- The meniscus needs to be set -only once- directly to the partial volume.
- Optimized flow geometry in the tip ensures reliable liquid dispensing from bulb and graduated pipettes.
- The mechanical resilience of the tip and its fire-polished edges ensure a long service life.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service