Skip to Content
Merck
All Photos(2)

Key Documents

Z2626

Sigma-Aldrich

Zaragozic acid A trisodium salt

from microbial, ≥95% (HPLC), lyophilized powder, squalene synthase inhibitor

Synonym(s):

L-694,599, Squalestatin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$691.00

$691.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$691.00

About This Item

Empirical Formula (Hill Notation):
C35H43O14Na3
CAS Number:
Molecular Weight:
756.68
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

$691.00


Check Cart for Availability

Request a Bulk Order

Product Name

Zaragozic acid A trisodium salt, ≥95% (HPLC), microbial

biological source

microbial

Quality Level

Assay

≥95% (HPLC)

form

lyophilized powder

solubility

DMSO: 9.80-10.20 mg/mL, clear, colorless to faintly yellow

shipped in

wet ice

storage temp.

−20°C

Application

Zaragozic acid A trisodium salt has been used as:
  • a component of serum-free medium for human embryonic kidney cells in a wound scratch assay to test its effect on wound healing[1]
  • a farnesyl-diphosphate farnesyltransferase 1 (FDFT1) or squalene synthase inhibitor in stromal cells[2]
  • a cholesterol synthesis inhibitor in T cells[3]

Biochem/physiol Actions

Potent inhibitor, in vivo and in vitro, of mammalian, fungal and Saccharomyces cervisiae squalene synthase. Squalene synthase is the first committed enzyme in sterol synthesis, catalyzing the reductive condensation of farnesyl pyrophosphate to form squalene. As a squalene synthase inhibitor, zaragozic acid produces lower plasma cholesterol levels in primates. Treatment of rats with zaragozic acid A caused an increase in hepatic low density lipoprotein (LDL) receptor mRNA levels.

Legal Information

Manufactured and sold under a restricted license from Merck & Co., Inc., Kenilworth, NJ, U.S.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isoprenoids increase bovine endometrial stromal cell tolerance to the cholesterol-dependent cytolysin from Trueperella pyogenes
Griffin S, et al.
Biology of Reproduction, 99(4), 749-760 (2018)
Topical mevastatin promotes wound healing by inhibiting the transcription factor c-Myc via the glucocorticoid receptor and the long non-coding RNA Gas5
Sawaya AP, et al.
Test, 293(4), 1439-1449 (2018)
OX40 complexes with phosphoinositide 3-kinase and protein kinase B (PKB) to augment TCR-dependent PKB signaling
So T, et al.
Journal of immunology (Baltimore, Md. : 1950), 186(6), 3547-3555 (2011)
Y Ichinose et al.
Gene, 80(2), 315-323 (1989-08-15)
We have isolated cDNA clones encoding the mouse cytokeratin No. 19 (Ck 19) from an intestinal cDNA library using synthetic oligodeoxyribonucleotides as probes. We obtained four independent clones, which correspond to about 1.4-kb of ck19 cDNA. Nucleotide sequence analysis revealed
Kaitlyn M Dykstra et al.
Oncotarget, 6(39), 41535-41549 (2015-11-26)
Multiple myeloma (MM) is characterized by the production of monoclonal protein (MP). We have shown previously that disruption of the isoprenoid biosynthetic pathway (IBP) causes a block in MP secretion through a disruption of Rab GTPase activity, leading to an

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service