Skip to Content
Merck
All Photos(1)

Key Documents

Y2021

Sigma-Aldrich

Yeast Synthetic Drop-out Medium Supplements

without histidine, leucine, tryptophan, and adenine

Synonym(s):

Yeast Synthetic Drop-out Media Supplements

Sign Into View Organizational & Contract Pricing

Select a Size

20 G
$189.00

$189.00


Available to ship on07 April 2025Details



Select a Size

Change View
20 G
$189.00

About This Item

MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

$189.00


Available to ship on07 April 2025Details


Quality Level

form

powder

technique(s)

transformation: suitable

application(s)

microbiology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

The selection of plasmids in yeast is based on the use of auxotrophic mutant strains that cannot grow without a specific media component (an amino acid, purine or pyrimidine). Transformation with a plasmid containing the mutated gene enables the transformant to grow on a medium lacking the required component. Sigma′s Yeast Synthetic Drop-Out Media Supplements create a richer medium for better yield and growth rate, and increase the probability of successful transformations when screening libraries or performing gene knock-outs.
Yeast Synthetic Drop-out Medium Supplements, without histidine, leucine, tryptophan, and adenine has been used in yeast two-hybrid screening.[1]

Other Notes

Mixtures of amino acids and other nutrients to be added to Yeast Nitrogen Base Without Amino Acids.

Preparation Note

Dissolve 1.39 g of supplement per liter of medium.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elham Alzyoud et al.
Frontiers in cell and developmental biology, 9, 727264-727264 (2021-10-19)
Microtubule nucleation in eukaryotes is primarily promoted by γ-tubulin and the evolutionary conserved protein complex, γ-Tubulin Ring Complex (γ-TuRC). γ-TuRC is part of the centrosome and basal body, which are the best-known microtubule-organizing centers. Centrosomes undergo intensive and dynamic changes
Corinne A Tovey et al.
Current biology : CB, 28(14), 2314-2323 (2018-07-10)
Microtubules are essential for various cell processes [1] and are nucleated by multi-protein γ-tubulin ring complexes (γ-TuRCs) at various microtubule organizing centers (MTOCs), including centrosomes [2-6]. Recruitment of γ-TuRCs to different MTOCs at different times influences microtubule array formation, but

Protocols

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Questions

  1. can I autoclave this particular yeast dropout medium supplement?

    1 answer
    1. Yes, this yeast drop-out medium supplement can be autoclaved. Please see the datasheet for additional preparation instructions:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/255/001/y2021dat.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service