Skip to Content
Merck
All Photos(4)

Key Documents

WH0084951M1

Sigma-Aldrich

Monoclonal Anti-TNS4 antibody produced in mouse

clone 3B8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CTEN, Anti-FLJ14950, Anti-PP14434, Anti-Tensin 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$548.00

$548.00


Check Cart for Availability


Select a Size

Change View
100 μG
$548.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$548.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNS4(84951)

General description

TNS4 (Tensin 4) is a signaling molecule localized in the prostate and placenta. It is a member of the TENSIN gene family. It has a Src homology 2 and phosphotyrosine-binding domains. Its expression has been observed in the early progression of melanoma cells.

Immunogen

TNS4 (AAH18706, 1 a.a. ~ 480 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGSKASSPHGLGSPLVASPRLEKRLGGLAPQRGSRISVLSASPVSDVSYMFGSSQSLLHSSNSSHQSSSRSLESPANSSSSLHSLGSVSLCTRPSDFQAPRNPTLTMGQPRTPHSPPLAKEHASSCPPSITNSMVDIPIVLINGCPEPGSSPPQRTPGHQNSVQPGAASPSNPCPATRSNSQTLSDAPFTTCPEGPARDMQPTMKFVMDTSKYWFKPNITREQAIELLRKEEPGAFVIRDSSSYRGSFGLALKVQEVPASAQSRPGEDSNDLIRHFLIESSAKGVHLKGADEEPYFGSLSAFVCQHSIMALALPCKLTIPQRELGGADGASDSTDSPASCQKKSAGCHTLYLSSVSVETLTGALAVQKAISTTFERDILPTPTVVHFKVTEQGITLTDVQRKVFFRRHYPLTTLRFCGMDPEQRKWQKYCKPSWIFGFVAKSQTEPQENVCHLFAEYDMVQPASQVIGLVTALLQDAERM

Biochem/physiol Actions

TNS4 (Tensin 4) is mainly involved in cell migration activity. It exhibits oncogenic property in colonic, breast, lung, and gastric cancers. In studies, its overexpression has been reported in the pancreatic cancer cells. In pancreatic cancer cells, TNS4 promotes colony formation and cell motility. It has been predicted that TNS4 could be a prognostic marker for primary melanoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su Hao Lo et al.
Cancer research, 62(15), 4217-4221 (2002-08-03)
Tensin is a signaling molecule that binds to actin filaments and localizes to focal adhesions. Recently, we have discovered that tensin represents a new gene family with related members that are expressed in a variety of tissues. In this study
Cecilia Sjoestroem et al.
PloS one, 8(11), e80492-e80492 (2013-11-19)
C-terminal tensin-like protein (Cten) is a focal adhesion protein originally identified as a tumor suppressor in prostate cancer. It has since been found to be overexpressed and function as an oncogene in numerous other cancers, but the expression status of
Saleh Al-Ghamdi et al.
Pancreas, 42(1), 135-140 (2012-07-04)
C-terminal tensin-like gene (CTEN, also known as TNS4) localizes to focal adhesions and is reported to function as an oncogene in colonic, breast, lung, and gastric cancers. Its role in pancreatic cancer is unknown and was thus investigated in this
Abdulkader Albasri et al.
Breast cancer research and treatment, 126(1), 47-54 (2010-04-15)
C-terminal tensin-like (CTEN) gene is a member of the TENSIN gene family, involved in cell migration and localised at focal adhesion sites. This study was designed to explore the prevalence and clinical significance of CTEN expression in a large and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service