Skip to Content
Merck
All Photos(6)

Documents

WH0084254M1

Sigma-Aldrich

Monoclonal Anti-CAMKK1 antibody produced in mouse

clone 1F6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CAMKKA, Anti-DKFZp761M0423, Anti-MGC34095, Anti-calcium/calmodulin-dependent protein kinase kinase 1, alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMKK1(84254)

General description

Calcium/calmodulin-dependent protein kinase kinase 1 (CAMKK1) is a member of the serine/threonine-protein kinase family. The CAMKK1 gene is mapped to human chromosome 17p13.2. It is expressed in the brain, as well as in the thymus and spleen.

Immunogen

CAMKK1 (AAH31647, 1 a.a. ~ 520 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEGGPAVCCQDPRAELVERVAAIDVTHLEEADGGPEPTRNGVDPPPRARAASVIPGSTSRLLPARPSLSARKLSLQERPAGSYLEAQAGPYATGPASHISPRAWRRPTIESHHVAISDAEDCVQLNQYKLQSEIGKGAYGVVRLAYNESEDRHYAMKVLSKKKLLKQYGFPRRPPPRGSQAAQGGPAKQLLPLERVYQEIAILKKLDHVNVVKLIEVLDDPAEDNLYLALQNQAQNIQLDSTNIAKPHSLLPSEQQDSGSTWAARSVFDLLRKGPVMEVPCDKPFSEEQARLYLRDVILGLEYLHCQKIVHRDIKPSNLLLGDDGHVKIADFGVSNQFEGNDAQLSSTAGTPAFMAPEAISDSGQSFSGKALDVWATGVTLYCFVYGKCPFIDDFILALHRKIKNEPVVFPEGPEISEELKDLILKMLDKNPETRIGVPDIKLHPWVTKNGEEPLPSEEEHCSVVEVTEEEVKNSVRLIPSWTTVILVKSMLRKRSFGNPFEPQARREERSMSAPGNLLV

Biochem/physiol Actions

Calcium/calmodulin-dependent protein kinase 1 (CAMKK1) participates in the calcium/calmodulin-dependent (CaM) kinase cascade. It phosphorylates CAMK1 at threonine 177. Functionally, the members of the CAMKKs family are involved in neurite development, long-term memory (LTM), and axon elongation.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Largo et al.
Haematologica, 92(6), 795-802 (2007-06-07)
Multiple myeloma (MM) is a malignant plasma cell neoplasia in which genetic studies have shown that genomic changes may affect almost all chromosomes, as shown by fluorescence in situ hybridization (FISH) and comparative genomic hybridization (CGH). Our objective was the
André da Silva Santiago et al.
Scientific reports, 8(1), 14800-14800 (2018-10-06)
The calcium/calmodulin-dependent protein kinases (CAMKKs) are upstream activators of CAMK1 and CAMK4 signalling and have important functions in neural development, maintenance and signalling, as well as in other aspects of biology such as Ca2+ signalling in the cardiovascular system. To
Kazutaka Nanba et al.
Endocrinology, 156(5), 1750-1756 (2015-02-14)
There is considerable evidence supporting the role of calcium signaling in adrenal regulation of both aldosterone synthase (CYP11B2) and aldosterone production. However, there have been no studies that investigated the role played by the Ca(2+)/calmodulin-dependent protein kinase kinase (CaMKK) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service