Skip to Content
Merck
All Photos(1)

Key Documents

WH0065008M1

Sigma-Aldrich

Monoclonal Anti-MRPL1 antibody produced in mouse

clone 2B7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BM022, Anti-L1mt, Anti-mitochondrial ribosomal protein L1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$484.00

$484.00


Check Cart for Availability


Select a Size

Change View
100 μG
$484.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$484.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRPL1(65008)

Related Categories

General description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the L1 ribosomal protein family. (provided by RefSeq)

Immunogen

MRPL1 (AAH15109, 1 a.a. ~ 303 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MVYQTSLCSCSVNIRVPNRHFAAAKKSAKKTKKGAKEKTPDEKKDEIEKIKAYPYMEGEPEDDVYLKRLYPRQIYEVEKAVHLLKKFQILDFTSPKQSVYLDLTLDMALGKKKNVEPFTSVLSLPYPFASEINKVAVFTENASEVKIAEENGAASAGGTSLIQKIWDDEIVADFYVAVPEIMPELNRLRKKLNKKYPKLSRNSIGRDIPKMLELFKNGHEIKVDEERENFLQTKIATLDMSSDQIAANLQAVINEVCRHRPLNLGPFVVRAFLRSSTSEGLLLKIDPLLPKEVKNEESEKEDA

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service