Skip to Content
Merck
All Photos(3)

Key Documents

WH0063891M1

Sigma-Aldrich

Monoclonal Anti-RNF123 antibody produced in mouse

clone 3F8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FLJ12565, Anti-FP1477, Anti-KPC1, Anti-ring finger protein 123

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$548.00

$548.00


Check Cart for Availability


Select a Size

Change View
100 μG
$548.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$548.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF123(63891)

General description

The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. (provided by RefSeq)

Immunogen

RNF123 (NP_071347, 1216 a.a. ~ 1314 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ADYISADELAQVEQMLAHLTSASAQAAAASLPTSEEDLCPICYAHPISAVFQPCGHKSCKACINQHLMNNKDCFFCKTTIVSVEDWEKGANTSTTSSAA

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richa Khanna et al.
The FEBS journal, 285(12), 2243-2262 (2018-04-21)
Lamins are key nuclear proteins which are important for maintaining nuclear structure and function. Mutations in lamins cause a spectrum of genetic diseases termed as laminopathies. RING finger containing E3 ubiquitin ligase, RNF123, is transcriptionally upregulated in cells expressing rod

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service