Skip to Content
Merck
All Photos(4)

Key Documents

WH0023327M4

Sigma-Aldrich

Monoclonal Anti-NEDD4L antibody produced in mouse

clone 1D2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-KIAA0439, Anti-RSP5, Anti-hNedd42, Anti-neural precursor cell expressed, developmentally down-regulated 4-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEDD4L(23327)

Related Categories

General description

The gene NEDD4L (neural precursor cell expressed, developmentally down-regulated 4-like) is mapped to human chromosome 18q21. The protein localizes in the cytoplasm.

Immunogen

NEDD4L (AAH32597, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MATGLGEPVYGLSEDEGESRILRVKVVSGIDLAKKDIFGASDPYVKLSLYVADENRELALVQTKTIKKTLNPKWNEEFYFRVNPSNHRLLFEVFDENRLT

Biochem/physiol Actions

NEDD4L (neural precursor cell expressed, developmentally down-regulated 4-like) is an ubiquitin ligase which is responsible for the ubiquitination and degradation of proteins. It causes ubiquitination of the TGF (transforming growth factor)-β membrane receptor, activated Smad (mothers against decapentaplegic homolog)-2/3 and inhibitory Smad7. NEDD4L-mediated ubiquitination of kidney epithelial sodium channels (ENaC) subunits plays an important role in hypertension control.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ran Zhao et al.
Nucleic acids research, 43(16), 7838-7849 (2015-07-02)
The expression of DNA damage-binding protein 2 (DDB2) has been linked to the prognosis of ovarian cancer and its underlying transcription regulatory function was proposed to contribute to the favorable treatment outcome. By applying gene microarray analysis, we discovered neural
Albert Escobedo et al.
Structure (London, England : 1993), 22(10), 1446-1457 (2014-10-09)
We investigated the mechanisms of activation and degradation of the E3 ubiquitin ligase Nedd4L combining the available biochemical information with complementary biophysical techniques. Using nuclear magnetic resonance spectroscopy, we identified that the C2 domain binds Ca(2+) and inositol 1,4,5-trisphosphate (IP3) using
Go Kuratomi et al.
The Biochemical journal, 386(Pt 3), 461-470 (2004-10-22)
Inhibitory Smad, Smad7, is a potent inhibitor of TGF-beta (transforming growth factor-beta) superfamily signalling. By binding to activated type I receptors, it prevents the activation of R-Smads (receptor-regulated Smads). To identify new components of the Smad pathway, we performed yeast
H Chen et al.
European journal of human genetics : EJHG, 9(12), 922-930 (2002-02-13)
The validation of full-length cDNA represents a crucial step in gene identification and subsequent functional analysis. In searching for candidate genes for bipolar disorder on chromosome 18q21, a novel gene homologous to NEDD4 (Neural precursor cells expressed developmentally down-regulated) was
Fredrick J Rosario et al.
Clinical science (London, England : 1979), 130(7), 499-512 (2015-11-27)
Changes in placental amino acid transfer directly contribute to altered fetal growth, which increases the risk for perinatal complications and predisposes for the development of obesity, diabetes and cardiovascular disease later in life. Placental amino acid transfer is critically dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service