Skip to Content
Merck
All Photos(5)

Key Documents

WH0009189M1

Sigma-Aldrich

Monoclonal Anti-ZBED1 antibody produced in mouse

clone 5G1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ALTE, Anti-KIAA0785, Anti-TRAMP, Anti-zinc finger, BED-type containing 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5G1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZBED1(9189)

General description

Zinc finger, BED-type containing 1 (ZBED1), also known as DNA replication-related element-binding factor (DREF), is encoded by the single-exon gene mapped to Xp/Yp PAR region that has a functional homolog on the Y chromosome and escapes X-inactivation.

Immunogen

ZBED1 (NP_004720, 3 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
NKSLESSQTDLKLVAHPRAKSKVWKYFGFDTNAEGCILQWKKIYCRICMAQIAYSGNTSNLSYHLEKNHPEEFCEFVKSNTEQMREAFATAFSKLKP

Biochem/physiol Actions

Zinc finger, BED-type containing 1 (ZBED1) function as a crucial transcription factor for cell proliferation, and is implicated in cell cycle-dependent regulation of a number of ribosomal protein (RP) genes. The encoded protein has been suggested to be involved in the transposition of other transposable elements.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Esposito et al.
Human molecular genetics, 8(1), 61-67 (1999-01-15)
We report the cloning of a novel gene, called Tramp, in the Xp/Yp PAR region that has a functional homologue on the Y chromosome and escapes X-inactivation. This gene encodes, within a single exon, a putative protein that has amino
A Helena Mangs et al.
Current genomics, 8(2), 129-136 (2008-07-29)
The pseudoautosomal regions (PAR1 and PAR2) of the human X and Y chromosomes pair and recombine during meiosis. Thus genes in this region are not inherited in a strictly sex-linked fashion. PAR1 is located at the terminal region of the
Sofie Traynor et al.
Nucleic acids research, 47(13), 6668-6684 (2019-05-23)
Rearrangement of the 1q12 pericentromeric heterochromatin and subsequent amplification of the 1q arm is commonly associated with cancer development and progression and may result from epigenetic deregulation. In many premalignant and malignant cells, loss of 1q12 satellite DNA methylation causes
Daisuke Yamashita et al.
Molecular and cellular biology, 27(6), 2003-2013 (2007-01-16)
Although ribosomal proteins (RPs) are essential cellular constituents in all living organisms, mechanisms underlying regulation of their gene expression in mammals remain unclear. We have established that 22 out of 79 human RP genes contain sequences similar to the human
Simone Johansen et al.
Genes, 13(1) (2022-01-22)
The transcription factor ZBED1 is highly expressed in trophoblast cells, but its functions in the processes of trophoblast and placental biology remain elusive. Here, we characterized the role of ZBED1 in trophoblast cell differentiation using an in vitro BeWo cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service