Skip to Content
Merck
All Photos(2)

Key Documents

WH0008277M1

Sigma-Aldrich

Monoclonal Anti-TKTL1 antibody produced in mouse

clone 1C10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TKR, Anti-TKT2, Anti-transketolase-like 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$484.00

$484.00


Estimated to ship on02 June 2025



Select a Size

Change View
100 μG
$484.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$484.00


Estimated to ship on02 June 2025


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C10, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TKTL1(8277)

Immunogen

TKTL1 (AAH25382, 1 a.a. ~ 596 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MADAEARAEFPEEARPDRGTLQVFQDMASRLRIHSIRATCSTSSGHPTSCSSSSEIMSVLFFYIMRYKQSDPENPDNDRFVLAKRLSFVDVATGWLGQGLGVACGMAYTGKYFDRASYRVFCLMSDGESSEGSVWEAMAFASYYSLDNLVATFDVNRLGHSGALPAEHCINIYQRRCEAFGWNTYVVDGRDVEALCQVFWQASQVKHKPTAVVAKTFKGRGTPSIEDAESWHAKPMPRERADAIIKLIESQIQTSRNLDPQPPIEDSPEVNITDVRMTSPPDYRVGDKIATRKACGLALAKLGYANNRVVVLDGDTRYSTFSEIFNKEYPERFIECFMAEQNMVSVALGCASRGRTIAFASTFAAFLTRAFDHIRIGGLAESNINIIGSHCGVSVGDDGASQMALEDIAMFRTIPKCTIFYPTDAVSTEHAVALAANAKGMCFIRTTRPETMVIYTPQERFEIGQAKVLRHCVSDKVTVIGAGITVYEALAAADELSKQDIFIRVIDLFTIKPLDVATIVSSAKATEGRIITVEDHYPQGGIGEAVCAAVSMDPDIQVHSLAVSGVPQSGKSEELLDMYGISARHIIVAVKCMLLN

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Rolland et al.
Human reproduction (Oxford, England), 28(1), 199-209 (2012-10-02)
Can protein biomarkers of the male genital tract be identified in human seminal plasma? We identified potential biomarkers for each of the organs participating in the secretions of the human seminal plasma. The seminal plasma fulfills critical functions for fertility

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service