Skip to Content
Merck
All Photos(2)

Key Documents

WH0000516M1

Sigma-Aldrich

Monoclonal Anti-ATP5G1 antibody produced in mouse

clone 1A12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9), Anti-ATP5A, Anti-ATP5G

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$645.00

$645.00


Estimated to ship on30 April 2025



Select a Size

Change View
100 μG
$645.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$645.00


Estimated to ship on30 April 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP5G1(516)

General description

ATP synthase membrane subunit c locus 1 (ATP5G1) is encoded by the gene mapped to human chromosome 17q21.32. The encoded protein belongs to the low transcript gene group.
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (α, β, γ, δ, and ε) assembled with a stoichiometry of 3 α, 3 β, and a single representative of the other 3. The proton channel seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene is one of three genes that encode subunit c of the proton channel. Each of the three genes have distinct mitochondrial import sequences but encode the identical mature protein. Alternatively spliced transcript variants encoding the same protein have been identified. (provided by RefSeq)

Immunogen

ATP5G1 (AAH04963, 18 a.a. ~ 136 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TRGLIRPVSASFLSSPVNSSKQPSYSNFPLQVARREFQTSVVSRDIDTAAKFIGAGAATVGVAGSGAGIGTVFGSLIIGYARNPSLKQQLFSYAILGFALSEAMGLFCLMVAFLILFAM

Biochem/physiol Actions

ATP synthase membrane subunit c locus 1 (ATP5G1) is one among the three genes that encodes mitochondrial ATP synthase subunit c of the proton channel. Regulation of this protein is implicated in the biogenesis of mammalian H+ -ATP synthase. Mutation in the gene is associated with the development of coronary artery disease (CAD).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

cDNA, genomic sequence cloning and overexpression of giant panda (Ailuropoda melanoleuca) mitochondrial ATP synthase ATP5G1.
Hou WR, et al.
Genetics and molecular research : GMR, 11(3), 3164-3174 (2012)
Evaluating the association of common UBE2Z variants with coronary artery disease in an Iranian population
Bastami M, et al.
Cellular and Molecular Biology, 61(7), 50-54 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service