Skip to Content
Merck
All Photos(8)

Key Documents

WH0000060M1

Sigma-Aldrich

Anti-β-Actin (ACTB) Antibody

mouse monoclonal, 3G4-F9

Synonym(s):

Anti-actin, beta

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$759.00

$759.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1312


Select a Size

Change View
100 μG
$759.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$759.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1312

Product Name

Monoclonal Anti-ACTB antibody produced in mouse, clone 3G4-F9, purified immunoglobulin, buffered aqueous solution

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G4-F9, monoclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTB(60)

General description

Actin β (ACTB) is encoded by the gene mapped to human chromosome 7p22.1. ACTB is a ubiquitously expressed cytoskeletal protein.
This gene encodes one of six different actin proteins. Actins are highly conserved proteins that are involved in cell motility, structure, and integrity. This actin is a major constituent of the contractile apparatus and one of the two nonmuscle cytoskeletal actins. (provided by RefSeq)

Immunogen

ACTB (AAH01301, 1 a.a. ~ 375 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDDDIAALVVDNGSGMCKAGFAGDDAPRAVSPSIVGRPRHQGVMVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIVTNWDDMEKIWHHTFYNELRVAPEEHPVLLTEAPLNPKANREKMTQIMFETFNTPAMYVAIQAVLSLYASGRTTGIVMDSGDGVTHTVPIYEGYALPHAILRLDLAGRDLTDYLMKILTERGYSFTTTAEREIVRDIKEKLCYVALDFEQEMATAASSSSLEKSYELPDGQVITIGNERFRCPEALFQPSFLGMESCGIHETTFNSIMKCDVDIRKDLYANTVLSGGTTMYPGIADRMQKEITALAPSTMKIKIIAPPERRYSVWIGGSILASLSTFQQMWISKQEYDESGPSIVHRKCF

Application

Monoclonal Anti-ACTB antibody produced in mouse has been used in western blots and immunohistochemistry.

Biochem/physiol Actions

Actin β (ACTB) is an endogenous housekeeping gene. It is extensively used as a reference gene/protein in quantifying expression levels in tumors. Mutation in the gene is associated with the development of Baraitser–Winter cerebrofrontofacial syndrome (BWCFF). Down regulated expression of the gene has been observed in liver, melanoma, renal, colorectal, gastric, pancreatic, esophageal, lung, breast, prostate, ovarian cancers, leukemia and lymphoma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?-Actin and GAPDH housekeeping gene expression in asthmatic airways is variable and not suitable for normalising mRNA levels
Glare EM
Thorax, 57, 765-770 (2002)
Breast Cancers Activate Stromal Fibroblast-Induced Suppression of Progenitors in Adjacent Normal Tissue.
Chatterjee S
Stem Cell Reports, 10, 196-211 (2018)
ACTB in cancer
Guo C
Clinica Chimica Acta; International Journal of Clinical Chemistry, 39-44 (2013)
Loss of Igfbp7 Causes Precocious Involution in Lactating Mouse Mammary Gland
Chatterjee S
PLoS ONE, 9 (2014)
Sumanta Chatterjee et al.
PloS one, 9(2), e87858-e87858 (2014-02-08)
Insulin like growth factors (IGFs) and their binding proteins (IGFBPs) are secreted peptides that play major roles in regulating the normal development and maturation of mammary gland. While Igfbp7 has been shown to decrease breast tumor growth, its role in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service