Skip to Content
Merck
All Photos(2)

Key Documents

V9125

Sigma-Aldrich

Snake venom from Naja naja kaouthia

Synonym(s):

Thailand Cobra

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
Product V9125 is not currently sold in your country. Contact Technical Service

storage temp.

−20°C

Application

Source of α-cobrotoxin, cardiotoxin and cobra venom anticomplementary protein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karina Juhl Rasmussen et al.
Journal of immunological methods, 444, 51-55 (2017-02-09)
The complement component C3 and the cleavage products of C3b/iC3b, C3c and C3d are used as biomarkers in clinical diagnostics. Currently, no specific antibodies are able to differentiate C3d from other fragments, although such a distinction could be very valuable

Questions

  1. What is the solubility of this product in water and or PBS?

    1 answer
    1. This product is not tested for solubility. However, cobra venom is reportedly soluble at 20 mg/mL in water to yield a clear to slightly hazy solution. Store the solution in aliquots - 20°C or -80°C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service