All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H20N2O3S
CAS Number:
Molecular Weight:
248.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
CSCCC(NC(=O)C(N)C(C)C)C(O)=O
InChI
1S/C10H20N2O3S/c1-6(2)8(11)9(13)12-7(10(14)15)4-5-16-3/h6-8H,4-5,11H2,1-3H3,(H,12,13)(H,14,15)
InChI key
YSGSDAIMSCVPHG-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
?OH radical induced decarboxylation of methionine-containing peptides. Influence of peptide sequence and net charge
Bobrowski K., Christian S, et al.
Journal of the Chemical Society, 5, 353-362 (1991)
Oxidation of methionine peptides by Fenton systems: the importance of peptide sequence, neighbouring groups and EDTA
Schoneich C. and Yang J.
Journal of the Chemical Society, 10, 915-924 (1996)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service