Skip to Content
Merck
All Photos(1)

Key Documents

V3890

Sigma-Aldrich

VU0240551

≥98% (HPLC)

Synonym(s):

CID 7211972, N-(4-Methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]-acetamide, SID 56405457, VU0240551-1, VU0240551-2-D4

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$192.00

$192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$192.00

About This Item

Empirical Formula (Hill Notation):
C16H14N4OS2
CAS Number:
Molecular Weight:
342.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$192.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

Cc1csc(NC(=O)CSc2ccc(nn2)-c3ccccc3)n1

InChI

1S/C16H14N4OS2/c1-11-9-23-16(17-11)18-14(21)10-22-15-8-7-13(19-20-15)12-5-3-2-4-6-12/h2-9H,10H2,1H3,(H,17,18,21)

InChI key

WJRWSLORVIHRNX-UHFFFAOYSA-N

Biochem/physiol Actions

VU0240551 is a potent, selective KCC2 inhibitor.
VU0240551 is a potent, selective KCC2 inhibitor. KCC2 is a potassium-chloride exchanger expressed specifically in neurons. KCC2 functions to lower intracellular chloride concentrations below the electrochemical potential of the cells, thereby increasing the hyperexcitability of the neurons. KCC2 activity enhances GABA and other inhibitory neurotransmission and is implicated in pain processing. VU0240551 was discovered in a high-throughput screen, followed by directed medicinal chemistry. VU0240551 is selective for KCC2 over NKCC1. VU0240551 binds competitively to the K+ site and binds noncompetitively to the Cl- site. VU0240551 is the only small molecule with specificity for a KCC family member.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mogens Andreasen et al.
Journal of neurophysiology, 117(4), 1512-1523 (2017-01-20)
The loop diuretic furosemide is known to have anticonvulsant effects, believed to be exerted through blockade of glial Na
Nicolas Doyon et al.
PLoS computational biology, 7(9), e1002149-e1002149 (2011-09-21)
Chloride homeostasis is a critical determinant of the strength and robustness of inhibition mediated by GABA(A) receptors (GABA(A)Rs). The impact of changes in steady state Cl(-) gradient is relatively straightforward to understand, but how dynamic interplay between Cl(-) influx, diffusion
Josiane C S Mapplebeck et al.
Cell reports, 28(3), 590-596 (2019-07-18)
The behavioral features of neuropathic pain are not sexually dimorphic despite sex differences in the underlying neuroimmune signaling. This raises questions about whether neural processing is comparably altered. Here, we test whether the K+-Cl- co-transporter KCC2, which regulates synaptic inhibition
Martin Heubl et al.
Nature communications, 8(1), 1776-1776 (2017-11-28)
The K+-Cl- co-transporter KCC2 (SLC12A5) tunes the efficacy of GABAA receptor-mediated transmission by regulating the intraneuronal chloride concentration [Cl-]i. KCC2 undergoes activity-dependent regulation in both physiological and pathological conditions. The regulation of KCC2 by synaptic excitation is well documented; however
Kwan Yeop Lee et al.
Pain, 156(12), 2431-2437 (2015-07-18)
Synaptic inhibition plays a key role in processing somatosensory information. Blocking inhibition at the spinal level is sufficient to produce mechanical allodynia, and many neuropathic pain conditions are associated with reduced inhibition. Disinhibition of spinal neurons can arise through decreased

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service