Skip to Content
Merck
All Photos(1)

Key Documents

V3375

Sigma-Aldrich

Bee venom from Apis mellifera (honey bee)

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

lyophilized powder

storage temp.

−20°C

Linkage

This preparation comes from a different supplier.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heera Yoon et al.
Biological & pharmaceutical bulletin, 38(5), 710-714 (2015-03-11)
Oxaliplatin, a platinum-based chemotherapy drug, often induces acute neuropathic pain, especially cold allodynia, even after a single administration. Subcutaneous injection of diluted bee venom (BV) into acupoints has been used to treat various pain symptoms in traditional oriental medicine. Although

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service