Skip to Content
Merck
All Photos(3)

Key Documents

V3264

Sigma-Aldrich

VDM 11

≥98% (HPLC), oil

Synonym(s):

(5Z,8Z,11Z,14Z)-N-(4-Hydroxy-2-methylphenyl)-5,8,11,14-eicosatetraenamide, N-(4-Hydroxy-2-methylphenyl)arachidonylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39NO2
CAS Number:
Molecular Weight:
409.60
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

description

Light-sensitive; store under argon.

Assay

≥98% (HPLC)

form

oil

color

gold

solubility

DMSO: soluble >5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)Nc1ccc(O)cc1C

InChI

1S/C27H39NO2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-27(30)28-26-22-21-25(29)23-24(26)2/h7-8,10-11,13-14,16-17,21-23,29H,3-6,9,12,15,18-20H2,1-2H3,(H,28,30)/b8-7-,11-10-,14-13-,17-16-

InChI key

WUZWFRWVRHLXHZ-ZKWNWVNESA-N

Biochem/physiol Actions

Potent, selective anandamide membrane transporter (AMT) inhibitor.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thiago Roberto Lima Romero et al.
Pharmacological reports : PR, 72(1), 96-103 (2020-02-06)
Xylazine is an α2 adrenoceptor agonist that is extensively used in veterinary medicine and animal experimentation procedures to produce analgesia, sedation and muscle relaxation without causing general anesthesia. Considering the lack of knowledge of the mechanisms involved in peripheral antinociception
L De Petrocellis et al.
FEBS letters, 483(1), 52-56 (2000-10-18)
Some synthetic agonists of the VR1 vanilloid (capsaicin) receptor also inhibit the facilitated transport into cells of the endogenous cannabinoid anandamide (arachidonoylethanolamide, AEA). Here we tested several AEA derivatives containing various derivatized phenyl groups or different alkyl chains as either
Yunbok Kim et al.
Frontiers in physiology, 13, 882176-882176 (2022-05-03)
Songbirds, such as zebra finches, spontaneously produce many song renditions for vocal practice even in the absence of apparent recipients throughout their lives. Such "undirected singing" is driven by intrinsic motivation, which arises within individuals for internal satisfaction without immediate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service