Skip to Content
Merck
All Photos(1)

Documents

V0883

Sigma-Aldrich

Val-Ala-Pro-Gly acetate salt

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26N4O5 · xC2H4O2
CAS Number:
Molecular Weight:
342.39 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)C(N)C(=O)NC(C)C(=O)N1CCCC1C(=O)NCC(O)=O

InChI

1S/C15H26N4O5/c1-8(2)12(16)14(23)18-9(3)15(24)19-6-4-5-10(19)13(22)17-7-11(20)21/h8-10,12H,4-7,16H2,1-3H3,(H,17,22)(H,18,23)(H,20,21)

InChI key

PPINMSZPTPRQQB-UHFFFAOYSA-N

Application

Serves as a quantitative marker for human elastins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea S Gobin et al.
Journal of biomedical materials research. Part A, 67(1), 255-259 (2003-10-01)
The elastin-derived peptide val-ala-pro-gly (VAPG) may be useful as a biospecific cell adhesion ligand for smooth muscle cells. By grafting the peptide sequence into a hydrogel material, we were able to assess its effects on smooth muscle cell adhesion and
L S Price et al.
Matrix (Stuttgart, Germany), 13(4), 307-311 (1993-07-01)
Thermolysin digests of human elastins were examined for reliable elastin peptide markers as determined by HPLC followed by amino acid sequencing of promising peaks. The tetrapeptide VAPG was found to occur in the early portion of the chromatogram in a
Peter Pocza et al.
International journal of cancer, 122(9), 1972-1980 (2007-12-14)
Melanomas containing more elastin are associated with higher stages of the disease. The interaction between elastin-derived peptides and melanoma cells appears to play an important role in the progression of melanomas. The effects of the elastin-derived peptides VGVAPG and VAPG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service