Skip to Content
Merck
All Photos(1)

Key Documents

UC602

Sigma-Aldrich

Phenyl-d5-7-hydroxywarfarin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$1,590.00
10 MG
$2,930.00

$1,590.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$1,590.00
10 MG
$2,930.00

About This Item

Empirical Formula (Hill Notation):
C19H11D5O5
CAS Number:
Molecular Weight:
329.36
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

$1,590.00


Estimated to ship on22 May 2025


Request a Bulk Order

form

solid

color

white

mp

201-203 °C

mass shift

M+5

storage temp.

2-8°C

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(CC(C)=O)C2=C(O)c3ccc(O)cc3OC2=O

InChI

1S/C19H16O5/c1-11(20)9-15(12-5-3-2-4-6-12)17-18(22)14-8-7-13(21)10-16(14)24-19(17)23/h2-8,10,15,21-22H,9H2,1H3/i2D,3D,4D,5D,6D

InChI key

SKFYEJMLNMTTJA-VIQYUKPQSA-N

Application

7-Hydroxywarfarin internal standard.

Biochem/physiol Actions

Labeled metabolite of warfarin.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mika Kosaka et al.
Drug metabolism and disposition: the biological fate of chemicals, 48(8), 655-661 (2020-06-03)
Pyrrolo[2,1-c][1,4]benzodiazepine dimer (PBD) has shown broad antitumor properties and potential as a therapeutic agent for cancers. During a routine drug-drug interaction assessment, it was found that PBD is a reversible inhibitor of CYP2C8 (IC50 = 1.1 µM) but not CYP1A2
Yoshiya Yamamura et al.
Aquatic toxicology (Amsterdam, Netherlands), 233, 105792-105792 (2021-03-05)
Although anticoagulant rodenticides (ARs) are effectively used for the control of invasive rodents, nontarget species are also frequently exposed to ARs and secondary poisonings occur widely. However, little data is available on the effects of ARs, especially on marine organisms.
Bianca M Liederer et al.
Xenobiotica; the fate of foreign compounds in biological systems, 49(9), 1063-1077 (2018-09-28)
GNE-617 (N-(4-((3,5-difluorophenyl)sulfonyl)benzyl)imidazo[1,2-a]pyridine-6-carboxamide) is a potent, selective nicotinamide phosphoribosyltransferase (NAMPT) inhibitor being explored as a potential treatment for human cancers. Plasma clearance was low in monkeys and dogs (9.14 mL min-1 kg-1 and 4.62 mL min-1 kg-1, respectively) and moderate in mice and rats (36.4 mL

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service