Skip to Content
Merck
All Photos(1)

Key Documents

U2508

Sigma-Aldrich

Ubiquitin Carrier Protein H2 human

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$451.00

$451.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$451.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

$451.00


Check Cart for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli (GST-tagged)

Assay

>95% (SDS-PAGE)

form

solution

mol wt

~14 kDa

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

Biochem/physiol Actions

UbcH2 is the human homolog to the yeast DNA repair gene RAD6 and is inducible by DNA damaging agents. Also associated with cancer-induced cachexia and the degradation of "N-end rule" substrates. UbcH2 mRNA level in muscle has also been correlated with sepsis. Moreover, c-Jun and c-Fos transcription factors have been shown to be ubiquitinated by an UbcH2-mediated event.
UbcH2 is the human homolog to the yeast DNA repair gene RAD6 and is inducible by DNA damaging agents. Also associated with cancer-induced cachexia and the degradation of "N-end rule" substrates. UbcH2 mRNA level in muscle has also been correlated with sepsis. Moreover, c-Jun and c-Fos transcription factors have been shown to be ubiquitinated by an UbcH2-mediated event. TNF-α was shown to stimulate expression of UbcH2 in skeletal muscle cells by increasing binding of NFκ-B to the promoter region of UbcH2, inducing transcription. [1]

Physical form

Solution in 50 mM HEPES, 50 mM NaCl, 10% glycerol, 1mM DTT, pH 7.8

Other Notes

manufactured for Sigma by Boston Biochem. Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Ping Li et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 17(9), 1048-1057 (2003-05-30)
In some inflammatory diseases, TNF-alpha is thought to stimulate muscle catabolism via an NF-kappaB-dependent process that increases ubiquitin conjugation to muscle proteins. The transcriptional mechanism of this response has not been determined. Here we studied the potential role of UbcH2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service