Skip to Content
Merck
All Photos(1)

Key Documents

TMO436110

Nunc® 96-well immuno plate

polystyrene, non-sterile, without lid, white, case of 80 ea

Synonym(s):

assay microplate, surface treated microplate

Sign Into View Organizational & Contract Pricing

Select a Size

80 EACH
$903.00

$903.00


Estimated to ship on01 June 2025



Select a Size

Change View
80 EACH
$903.00

About This Item

UNSPSC Code:
41122102
NACRES:
NB.15

$903.00


Estimated to ship on01 June 2025


material

polystyrene
white

sterility

non-sterile

feature

reusable

packaging

case of 80 ea

manufacturer/tradename

Nunc 436110

binding type

hydrophillic surface

Looking for similar products? Visit Product Comparison Guide

General description

Thermo Scientific MaxiSorp is hydrophilic. The well is white in color.
Thermo Scientific White 96-Well Immuno Plates are used to perform both quantitative and qualitative immunoassays, such as ELISA and binding assays. It is designed to be compatible with commonly used instruments. These plates ensure excellent performance, consistent results between different lots, and reproducibility across individual wells.

Application

Nunc® 96-well immuno plate is suitable for antibody sandwich assays.

Features and Benefits

  • SBS footprint for compatibility with standard equipment
  • Wells are available in flat or round bottom configurations
  • Three surface types to optimize binding
  • Certified binding homogeneity/reproducibility
  • White color provides maximum reflection
  • Minimal autofluorescence and autoluminescence

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service