Skip to Content
Merck
All Photos(1)

Documents

TMO312189-0008

Nalgene® wide-mouth bottle

high-density polyethylene, capacity 250 mL, natural, case of 250 ea

Synonym(s):

chemically resistant bottle, packaging bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

high-density polyethylene

packaging

case of 250 ea

manufacturer/tradename

Nalgene 312189-0008

capacity

250 mL

color

natural

Looking for similar products? Visit Product Comparison Guide

General description

Confidently ship and store these Thermo Scientific Nalgene Wide-Mouth HDPE Packaging Bottles, which are easy to fill and resistant to chemicals. These wide-mouthed, high-density polyethelyene bottles are injection blow-molded for strength and uniform wall thickness.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service