Skip to Content
Merck
All Photos(1)

Key Documents

TMO312104-0004

Nalgene® wide-mouth bottle

high-density polyethylene, capacity 125 mL, natural, case of 500 ea

Synonym(s):

Nalgene bottle, plastic bottle

Sign Into View Organizational & Contract Pricing

Select a Size

500 EACH
$1,830.00

$1,830.00


Check Cart for Availability


Select a Size

Change View
500 EACH
$1,830.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

$1,830.00


Check Cart for Availability

material

high-density polyethylene

packaging

case of 500 ea

manufacturer/tradename

Nalgene 312104-0004

capacity

125 mL

color

natural

Looking for similar products? Visit Product Comparison Guide

General description

Confidently ship and store these Thermo Scientific Nalgene Wide-Mouth HDPE Packaging Bottles, which are easy to fill and resistant to chemicals. These wide-mouthed, high-density polyethelyene bottles are injection blow-molded for strength and uniform wall thickness.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service