Skip to Content
Merck
All Photos(1)

Key Documents

TMO3103-0050

Nalgene® centrifuge tubes

conical bottom clear polypropylene copolymer, volume 50 mL, without cap, non-sterile, case of 100 ea

Synonym(s):

Graduated Conical Bottom Centrifuge Tubes

Sign Into View Organizational & Contract Pricing

Select a Size

100 EACH
$1,650.00

$1,650.00


Check Cart for Availability


Select a Size

Change View
100 EACH
$1,650.00

About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

$1,650.00


Check Cart for Availability

material

conical bottom clear polypropylene copolymer

sterility

non-sterile

feature

without cap

packaging

case of 100 ea

manufacturer/tradename

Nalgene 3103-0050

volume

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

Simplify measurements using Thermo Scientific Nalgene PPCO Conical-Bottom Centrifuge Tubes, which feature molded-in graduations. Translucent polypropylene copolymer (PPCO) has excellent chemical resistance.

<UL><LI>Translucent Polypropylene Copolymer (PPCO) is compatible with a wide range of lab reagents</LI><LI>Designed for low-speed centrifugation up to 6000 × g in refrigerated or non-refrigerated centrifuges (Tubes must be filled at least 80% for proper performance)</LI><LI>Conical bottoms concentrate pellet in a small area for easy isolation and retrieval</LI><LI>Molded-in graduations last the life of the tube</LI><LI>Last longer than polycarbonate tubes under conditions of repeated autoclaving</LI><LI>Tube are both autoclavable and graduated</LI></UL>

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service