Skip to Content
Merck
All Photos(1)

Key Documents

T9549

Sigma-Aldrich

Thrombin from bovine plasma

powder, suitable for cell culture, ≥1,500 NIH units/mg protein (E1%/280 = 19.5)

Synonym(s):

Factor IIa

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

powder

specific activity

≥1,500 NIH units/mg protein (E1%/280 = 19.5)

mol wt

heavy chain ~31 kDa
light chain ~6 kDa

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

cow ... F2(280685)

Looking for similar products? Visit Product Comparison Guide

General description

Thrombin is the final coagulation protease in regard to hemostasis, promoting both procoagulant and anticoagulant effects.

Application

Thrombin is used for site specific cleavage of recombinant fusion proteins containing an accessible thrombin recognition site for removal of affinity tags. Thrombin has been used in a study to assess an expression and purification system for the biosynthesis of adenosine receptor peptides for biophysical and structural characterization.

Biochem/physiol Actions

Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

Unit Definition

Activity is expressed in NIH units, and is measured by direct comparison to an NIH thrombin reference standard.

Reconstitution

When reconstituted with 1 mL water, vial contains stated activity in 0.15 M sodium chloride and 0.05 M sodium citrate, pH 6.5.

Analysis Note

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard, Lot K.
The NIH assay procedure uses 0.2 mL of diluted plasma (1:1 with saline) as a substrate and 0.1 mL of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Other Notes

View more information on thrombin at www.sigma-aldrich.com/enzymeexplorer.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachary T Britton et al.
Protein expression and purification, 84(2), 224-235 (2012-06-23)
Biophysical and structural characterization of G protein-coupled receptors (GPCRs) has been limited due to difficulties in expression, purification, and vitro stability of the full-length receptors. "Divide and conquer" approaches aimed at the NMR characterization of peptides corresponding to specific regions
Andrew F Uehlin et al.
Pharmaceutics, 14(2) (2022-02-27)
In this study, we report a biohybrid oriented fibrous scaffold based on nanofibers of poly(l-lactic acid) (PLLA)/fibrin produced by electrospinning and subsequent post-treatment. Induced hydrolytic degradation of the fibers in 0.25 M NaOH solution for various time periods followed by
Yuya Nakatani et al.
Archives of oral biology, 73, 172-178 (2016-10-25)
Platelet-rich plasma (PRP) is typically isolated and applied immediately after preparation, making it both a time- and labor-intensive addition to the operative procedure. Thus, it would be convenient if PRP could be preserved. We evaluated the efficacy of freeze-dried PRP
Sharon Wei Ling Lee et al.
Advanced healthcare materials, 9(7), e1901486-e1901486 (2020-03-04)
Polymer nanoparticles (NPs), due to their small size and surface functionalization potential have demonstrated effective drug transport across the blood-brain-barrier (BBB). Currently, the lack of in vitro BBB models that closely recapitulate complex human brain microenvironments contributes to high failure
Walter-Michael Halbmayer et al.
Clinical chemistry and laboratory medicine, 50(9), 1601-1605 (2012-10-27)
Dabigatran etexilate is a new oral anticoagulant for the therapy and prophylaxis of venous thromboembolism and stroke prevention in patients with atrial fibrillation. To investigate the extent of interactions of this new anticoagulant with frequently used coagulation assays, we completed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service