T9505
L-Tyrosine benzyl ester p-toluenesulfonate salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H17NO3 · C7H8O3S
CAS Number:
Molecular Weight:
443.51
Beilstein:
3855339
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥97% (TLC)
form
powder
color
white
storage temp.
−20°C
SMILES string
Cc1ccc(cc1)S(O)(=O)=O.N[C@@H](Cc2ccc(O)cc2)C(=O)OCc3ccccc3
InChI
1S/C16H17NO3.C7H8O3S/c17-15(10-12-6-8-14(18)9-7-12)16(19)20-11-13-4-2-1-3-5-13;1-6-2-4-7(5-3-6)11(8,9)10/h1-9,15,18H,10-11,17H2;2-5H,1H3,(H,8,9,10)/t15-;/m0./s1
InChI key
PJGVHBLZZQDFFM-RSAXXLAASA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J Negrel et al.
European journal of biochemistry, 247(3), 1127-1135 (1997-08-01)
We report the purification of hydroxycinnamoyl-CoA:tyramine N-(hydroxycinnamoyl)transferase (THT) to apparent homogeneity in 12% yield from tobacco (Nicotiana tabacum L. cv. Xanthi) cell-suspension cultures elicited with a commercial preparation of pronase. The purification procedure employs only four chromatography steps and takes
Sangkyu Park et al.
Biotechnology letters, 31(6), 911-915 (2009-02-21)
The plant-specific tyramine derivatives, feruloyltyramine (FT) and 4-coumaroyltyramine (CT), represent bioactive compounds found at low levels in many plant species. We generated transgenic rice seeds that produce high levels of CT (14 microg g(-1) seeds) and FT (2.7 microg g(-1)
J Brtko et al.
Endocrine regulations, 26(3), 127-131 (1992-09-01)
The effect of protease inhibitors N-tosyl-L-phenylalanine chloromethyl ketone (TPCK) and N-carbobenzoxy-L-phenylalanine chloromethyl ketone (ZPCK) at concentrations ranging from 1.5 x 10(-6) mol/l to 1.5 x 10(-4) mol/l on the specific binding of 3,5,3'-triiodothyronine (T3) to rat liver nuclear receptors was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service