Skip to Content
Merck
All Photos(2)

Key Documents

T9269

Sigma-Aldrich

TAPSO

≥99% (titration)

Synonym(s):

2-Hydroxy-3-[tris(hydroxymethyl)methylamino]-1-propanesulfonic acid, N-[Tris(hydroxymethyl)methyl]-3-amino-2-hydroxypropanesulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$113.00

$113.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$113.00

About This Item

Empirical Formula (Hill Notation):
C7H17NO7S
CAS Number:
Molecular Weight:
259.28
Beilstein:
2113949
EC Number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

$113.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99% (titration)

form

crystalline powder

useful pH range

7.0-8.2

pKa (25 °C)

7.6

solubility

warm water: 0.333 g/mL, clear to slightly hazy, colorless

application(s)

diagnostic assay manufacturing

SMILES string

OCC(CO)(CO)NCC(O)CS(O)(=O)=O

InChI

1S/C7H17NO7S/c9-3-7(4-10,5-11)8-1-6(12)2-16(13,14)15/h6,8-12H,1-5H2,(H,13,14,15)

InChI key

RZQXOGQSPBYUKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Interpretation of non-Nernstian slopes in graphic analysis of data collected in pH range close to deprotonation of a ligand Part I. A glass electrode potentiometric and polarographic study of Cd-(TAPSO)x-(OH)y and Zn-(TAPSO)x-(OH)y systems.: Explores the complex behavior of TAPSO in metal binding studies, particularly with cadmium and zinc, providing insight into its utility in heavy metal chelation and environmental cleanup strategies (Machado et al., 2006).

  • Challenges in modelling and optimisation of stability constants in the study of Cu-(TAPS)(x)-(OH)(y) system by polarography.: Discusses the challenges in modeling and optimizing the stability constants for copper complexes with TAPSO, contributing to better understanding and utilization of this buffer in copper-related biochemical research (Machado and Soares, 2007).

  • Modelling of Pb-(TAPS)(x)-(OH)(y) system and refinement of stability constants in the region of lead hydrolysis and lead hydroxide precipitation.: Provides a detailed examination of TAPSO′s role in modeling lead complexation systems, which is critical for environmental monitoring and remediation efforts involving lead (Machado et al., 2007).

  • Synthetic organic pH buffers can support fertilization of guinea pig eggs, but not as efficiently as bicarbonate buffer.: Evaluates the efficacy of synthetic buffers like TAPSO in supporting fertilization, offering valuable data for reproductive biology and assisted reproductive technologies (Bhattacharyya and Yanagimachi, 1988).

Caution

According to Ferreira, et al., TAPSO can have a strong interaction with iron in solution.[1] Based on this citation, it is recommended to use plastic-coated or PTFE-coated utensils when working with this chemical.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Un)suitability of the use of pH buffers in biological, biochemical and environmental studies and their interaction with metal ions - a review.
Ferreira, C.M.H., et al.
Royal Society of Chemistry Advances, 5, 30989-31003 (2015)
(Un)suitability of the use of pH buffers in biological, biochemical and environmental studies and their interaction with metal ions ? a review.
Ferreira, C.M.H., et al.
Royal Society of Chemistry Advances, 5, 30989-31003 (2015)

Related Content

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service