Skip to Content
Merck
All Photos(1)

Key Documents

T8806

Sigma-Aldrich

Thymopentin

≥97% (HPLC)

Synonym(s):

TP5, Arg-Lys-Asp-Val-Tyr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H49N9O9
CAS Number:
Molecular Weight:
679.76
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Thymopentin, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white

application(s)

cell analysis

storage temp.

−20°C

SMILES string

CC(C)[C@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)CCCNC(N)=N)C(=O)N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C30H49N9O9/c1-16(2)24(28(46)38-22(29(47)48)14-17-8-10-18(40)11-9-17)39-27(45)21(15-23(41)42)37-26(44)20(7-3-4-12-31)36-25(43)19(32)6-5-13-35-30(33)34/h8-11,16,19-22,24,40H,3-7,12-15,31-32H2,1-2H3,(H,36,43)(H,37,44)(H,38,46)(H,39,45)(H,41,42)(H,47,48)(H4,33,34,35)/t19-,20-,21-,22-,24-/m0/s1

InChI key

PSWFFKRAVBDQEG-YGQNSOCVSA-N

Amino Acid Sequence

Arg-Lys-Asp-Val-Tyr

General description

Thymopentin (TP5) is a pentapeptide produced by the epithelial cells of the thymic cortex and medulla. It consists of thymosin α1 (Tα1).

Biochem/physiol Actions

Thymopentin (TP5) is an immunomodulator, resembling the active site of the thymopoietin. TP5 stimulates cellular immune responses such as T-cell formation, natural killer (NK) cell and macrophage activation, generation of cytokines, and activation of B-cell for antibody production. It also mimics the polypeptide environment of the thymus. TP5 is used to treat conditions like immune deficiency, autoimmune diseases, infections, and cancers. It has a short half-life and is rapidly metabolized in the gastrointestinal tract.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonia Patruno et al.
Nitric oxide : biology and chemistry, 27(3), 143-149 (2012-06-23)
While thymopentin has been used for many years in the experimental treatment of Sézary syndrome (SS), a rare and very aggressive lymphoma, its mechanism of action is still not known. Herein we show that this peptide acts as an inhibitor
Wendy B London et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 29(24), 3286-3292 (2011-07-20)
Survival after neuroblastoma relapse is poor. Understanding the relationship between clinical and biologic features and outcome after relapse may help in selection of optimal therapy. Our aim was to determine which factors were significantly predictive of postrelapse overall survival (OS)
Bo Yang et al.
Hematological oncology, 30(3), 115-122 (2012-09-14)
The elderly population is susceptible to haematological malignancies, and these elderly patients are intolerant to cytotoxic drugs. Therefore, the exploration of a safe and reliable strategy exclusive of chemotherapy is critical in improving the prognosis of elderly patients with haematological
G Goldstein et al.
Survey of immunologic research, 4 Suppl 1, 1-10 (1985-01-01)
Thymopoietin is a polypeptide hormone of the thymus consisting of 49 amino acids. The pentapeptide thymopentin (TP-5) Arg-Lys-Asp-Val-Tyr, corresponding to amino acids 32-36 of thymopoietin, appears to represent the active site of thymopoietin in that it has all the biological
Yan Zhang et al.
International journal of pharmaceutics, 386(1-2), 15-22 (2009-11-10)
Copolymers were synthesized by ring opening polymerization of l- or d-lactide in the presence of dihydroxyl PEG with molar mass of 6000, 12,000 and 20,000, using zinc lactate as catalyst. Bioresorbable hydrogels were obtained by mixing PLLA-PEG-PLLA and PDLA-PEG-PDLA aqueous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service