Skip to Content
Merck
All Photos(1)

Key Documents

T7687

Sigma-Aldrich

Anti-Tissue Inhibitor of Metalloproteinase-3, First Loop antibody produced in rabbit

~1 mg/mL, buffered aqueous glycerol solution, affinity isolated antibody

Synonym(s):

Anti-TIMP-3

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
$661.00

$661.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
0.1 MG
$661.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$661.00


Estimated to ship on16 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000 using a whole cell extract from human fibroblasts (M 3303)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TIMP3(7078)

Specificity

By immunoblotting against the reduced protein, the antibody reacts with bands at ~24 kDa and 30 kDa (unglycosylated and glycosylated, respectively). It also reacts with non-reduced TIMP-3.

Immunogen

synthetic peptide corresponding to the first loop of human tissue inhibitor of metalloproteinase-3.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 50% glycerol and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Saihan et al.
Molecular vision, 15, 1218-1230 (2009-06-19)
To determine the phenotypic and biochemical characteristics of the p.E139K missense variant in tissue inhibitor of metalloproteinase 3 (TIMP3) associated with Sorsby fundus dystrophy (SFD). The coding regions and adjacent intronic sequence of TIMP3 were amplified by polymerase chain reaction
Sarah Naessens et al.
Human mutation, 40(5), 539-551 (2019-01-23)
Sorsby fundus dystrophy (SFD) is a macular degeneration caused by mutations in TIMP3, the majority of which introduce a novel cysteine. However, the exact molecular mechanisms underlying SFD remain unknown. We aimed to provide novel insights into the functional consequences

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service