Skip to Content
Merck
All Photos(1)

Key Documents

T6829

Sigma-Aldrich

TUG-424

≥98% (HPLC)

Synonym(s):

3-(4-(o-Tolylethynyl)phenyl)propanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$233.00

$233.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$233.00

About This Item

Empirical Formula (Hill Notation):
C18H16O2
CAS Number:
Molecular Weight:
264.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$233.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

Cc1ccccc1C#Cc2ccc(CCC(O)=O)cc2

InChI

1S/C18H16O2/c1-14-4-2-3-5-17(14)12-10-15-6-8-16(9-7-15)11-13-18(19)20/h2-9H,11,13H2,1H3,(H,19,20)

InChI key

FODHWOBAQBTTFS-UHFFFAOYSA-N

Biochem/physiol Actions

TUG-424 is expected to be useful in the exploration of FFA1 and may also be valuable as a lead structure for new potential antidiabetic therapeutics.
TUG-424 is expected to be useful in the exploration of FFA1 and may also be valuable as a lead structure for new potential antidiabetic therapeutics. TUG-424 significantly increased glucose-stimulated insulin secretion at 100 nM and may serve to explore the role of FFA1 in metabolic diseases such as diabetes or obesity. It enhanced glucose-stimulated insulin secretion in a rat beta-cell line already at 100 nM and from isolated mouse islets through FFA1.

GPR40 (now FFA1) was formerly an orphan GPCR whose endogenous ligands have recently been identified as free fatty acids (FFAs). The receptor appears to be involved in the pathophysiology of type 2 diabetes and is a drug target because of its role in FFA-mediated enhancement of glucose-stimulated insulin release.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Luo et al.
PloS one, 7(10), e46300-e46300 (2012-10-12)
Type 2 diabetes is characterized by impaired glucose homeostasis due to defects in insulin secretion, insulin resistance and the incretin response. GPR40 (FFAR1 or FFA1) is a G-protein-coupled receptor (GPCR), primarily expressed in insulin-producing pancreatic β-cells and incretin-producing enteroendocrine cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service