Skip to Content
Merck
All Photos(1)

Key Documents

T6699

Sigma-Aldrich

pPolh-MAT-Tag®-1 Transfer Vector

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$754.00

$754.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
20 μG
$754.00

About This Item

UNSPSC Code:
12171500
NACRES:
NA.51

$754.00


Estimated to ship on02 June 2025


Request a Bulk Order

shipped in

dry ice

storage temp.

−20°C

Application

Transfer vector for generation of recombinant Baculovirus and expression of N-terminal MAT-Tag® fusions in insect cells.

Legal Information

MAT-Tag is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service