Skip to Content
Merck
All Photos(2)

Key Documents

T6412

Sigma-Aldrich

2,3,4,6-Tetra-O-benzyl-D-galactose

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H36O6
CAS Number:
Molecular Weight:
540.65
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥98%

form

powder

optical activity

[α]25/D 9.0 to 13.0 °, c = 1% (w/v) in chloroform

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

−20°C

SMILES string

OC1O[C@H](COCc2ccccc2)[C@H](OCc3ccccc3)[C@H](OCc4ccccc4)[C@H]1OCc5ccccc5

InChI

1S/C34H36O6/c35-34-33(39-24-29-19-11-4-12-20-29)32(38-23-28-17-9-3-10-18-28)31(37-22-27-15-7-2-8-16-27)30(40-34)25-36-21-26-13-5-1-6-14-26/h1-20,30-35H,21-25H2/t30-,31+,32+,33-,34?/m1/s1

InChI key

OGOMAWHSXRDAKZ-BJPULKCASA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service