Skip to Content
Merck
All Photos(1)

Documents

T5694

Sigma-Aldrich

TRAIL human

recombinant, expressed in NSO cells, >97% (SDS-PAGE), lyophilized powder

Synonym(s):

Apo2 Ligand, Apo2L, TL2, TNF-related apoptosis-inducing ligand

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in NSO cells

description

N-terminal 6X histidine-tagged extracellular domain

Assay

>97% (SDS-PAGE)

form

lyophilized powder

mol wt

24 kDa by SDS-PAGE
predicted mol wt ~21 kDa

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TNFSF10(8743)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

TRAIL human was used to induce cytotoxicity in A549, a human lung carcinoma cell line.

Biochem/physiol Actions

TNF-related apoptosis-inducing ligand (TRAIL) is a type II transmembrane protein that belongs to TNF family. It is present in both soluble as well as membrane-bound form and can induce rapid apoptosis of various transformed cell lines but not the normal cells. Additionally, the product can also induces apoptotic cell death in TRAIL-sensitive U343MGcells.
Type II transmembrane protein of the TNF superfamily. Ligand for apoptosis-inducing death receptors DR4 and DR5, decoy receptors DcR1 and DcR2, and osteoprotegerin. Approximately 28% amino acid sequence identity to FAS ligand. Human and mouse TRAIL share 65% sequence identity.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.5 mg bovine serum albumin.

Analysis Note

Induces cytotoxicity in mouse L-929 cells in the presence of actinomycin D.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Sträter et al.
Annals of the New York Academy of Sciences, 915, 162-170 (2001-02-24)
The tumor necrosis factor receptor (TNFR) family is a still-growing group of homologous transmembrane proteins, some of which bear an intracellular "death domain" and are able to directly mediate apoptosis. Apoptosis is induced upon trimerization of the receptors by their
Bich Hang Do et al.
Journal of microbiology and biotechnology, 27(12), 2156-2164 (2017-10-17)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is considered as an antitumor agent owing to its ability to induce apoptosis of cancer cells without imparting toxicity toward most normal cells. TRAIL is produced in poor yield because of its insoluble expression
X D Zhang et al.
FEBS letters, 482(3), 193-199 (2000-10-12)
Resistance of normal cells to tumour necrosis factor related apoptosis inducing ligand (TRAIL) induced apoptosis is believed to be mediated by expression of two decoy receptors. Here we show that the expression and localisation of TRAIL receptors (TRAIL-Rs) vary between
S R Wiley et al.
Immunity, 3(6), 673-682 (1995-12-01)
A novel tumor necrosis factor (TNF) family member has been cloned and characterized. This protein, designated TNF-related apoptosis-inducing ligand (TRAIL), consists of 281 and 291 aa in the human and murine forms, respectively, which share 65% aa identity. TRAIL is
W S Simonet et al.
Cell, 89(2), 309-319 (1997-04-18)
A novel secreted glycoprotein that regulates bone resorption has been identified. The protein, termed Osteoprotegerin (OPG), is a novel member of the TNF receptor superfamily. In vivo, hepatic expression of OPG in transgenic mice results in a profound yet nonlethal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service