Skip to Content
Merck
All Photos(1)

Key Documents

T5515

Sigma-Aldrich

Thionicotinamide adenine dinucleotidephosphate sodium salt

≥95%

Synonym(s):

Thionicotinamid-TPN sodium salt, Thionicotinamide-TPN, Triphosphothiopyridine nucleotide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H27N7O16P3S · Na
CAS Number:
Molecular Weight:
781.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95%

solubility

H2O: soluble 50 mg/mL, clear, yellow to very deep yellow

storage temp.

−20°C

SMILES string

[Na+].NC(=S)c1ccc[n+](c1)[C@@H]2O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]3O[C@H]([C@H](OP(O)([O-])=O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

InChI

1S/C21H28N7O16P3S.Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(43-45(32,33)34)14(30)11(42-21)6-40-47(37,38)44-46(35,36)39-5-10-13(29)15(31)20(41-10)27-3-1-2-9(4-27)18(23)48;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,48);/q;+1/p-1/t10-,11-,13-,14-,15-,16-,20-,21-;/m1./s1

InChI key

RFGBSJGYGUYCBT-QYZPTAICSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Thio-NADP has been identified as a compound unique to the β-arrestin CNR2 screening assay4.

Biochem/physiol Actions

Blocks nicotinate adenine dinucleotide phosphate (NAADP)-induced Ca2+ release
Studies using sea-urchin egg homogenates have reported that thio-NADP stimulates partial Ca2+ release at 50 μm and blocks successive challenge with a saturating concentration of nicotinic acid adenine dinucleotide phosphate (NAADP). Subsequent HPLC purification analysis have reported that Ca2+ release function of thio-NADP is because of the presence of NAADP as a contaminant. Hence, this study challenges the NAADP antagonist functions of thio-NADP and indicates that thio-NADP does not function as an NAADP blocker5.

Linkage

Analog of NADP

Preparation Note

Thionicotinamide adenine dinucleotidephosphate (thio-NADP) sodium is soluble in water at 50 mg/ml and yields a clear, yellow to dark yellow solution.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arkady S Pivovarov et al.
Regulatory peptides, 111(1-3), 31-39 (2003-03-01)
The effect of seven compounds intracellularly applied by spontaneous diffusion were investigated on the EPYLRFamide-induced reduction of acetylcholine-induced inward current (ACh-current) recorded from identified neurones from Helix lucorum. Inward currents were recorded from neurones LPa2, LPa3, RPa3 and RPa2 in
J Feeney et al.
Biochemistry, 22(3), 628-633 (1983-02-01)
Transferred nuclear Overhauser effect measurements have been made on complexes of NADP+ and thioNADP+ with Lactobacillus casei dihydrofolate reductase to provide information about the glycosidic bond conformations in these complexes. Both NADP+ and thioNADP+ are shown to have very similar
Hans-Gert Korth et al.
Biochemistry, 51(23), 4693-4703 (2012-05-24)
Mammalian (Clade 3) catalases utilize NADPH as a protective cofactor to prevent one-electron reduction of the central reactive intermediate Compound I (Cpd I) to the catalytically inactive Compound II (Cpd II) species by re-reduction of Cpd I to the enzyme's
D M Dickey et al.
Cell biochemistry and biophysics, 28(1), 63-73 (1997-12-05)
Nicotinic acid adenine dinucleotide phosphate (NAADP) is a metabolite of NADP, which can release Ca2+ from stores that are distinct from those activated by either cyclic ADP-ribose or inositol 1,4,5-trisphosphate (IP3). It has previously been suggested that thio-NADP is a
E N Chini et al.
The Biochemical journal, 316 ( Pt 3), 709-711 (1996-06-15)
We investigated the dependence of nicotinate-adenine dinucleotide phosphate (NAADP)-induced Ca2+ release from intracellular stores of sea urchin egg homogenates, upon extravesicular Ca2+. In contrast to the Ca2+ release induced inositol 1',4',5'-triphosphate (IP3) or cyclic ADP-ribose (cADPR), the Ca2+ release induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service