Skip to Content
Merck
All Photos(1)

Key Documents

T5406

Sigma-Aldrich

2,2′:5′,2′′-Terthiophene

Synonym(s):

α-Terthienyl, 2,5-Di(2-thienyl)thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8S3
CAS Number:
Molecular Weight:
248.39
Beilstein:
178604
MDL number:
UNSPSC Code:
12352200

mp

93-95 °C (lit.)

SMILES string

c1csc(c1)-c2ccc(s2)-c3cccs3

InChI

1S/C12H8S3/c1-3-9(13-7-1)11-5-6-12(15-11)10-4-2-8-14-10/h1-8H

InChI key

KXSFECAJUBPPFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Natural product occurring in marigold which shows UV-enhanced antibiotic activity.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service