T4168
Nalgene® centrifuge tubes, Round Style 3117
nominal capacity 16 mL
Synonym(s):
High-Speed Centrifuge Tubes, Nalgene tubes
Select a Size
$125.00
Estimated to ship on31 May 2025
Select a Size
About This Item
$125.00
Estimated to ship on31 May 2025
Recommended Products
material
polycarbonate
unlipped
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10
manufacturer/tradename
Nalgene® 3117-0160
nominal capacity
16 mL
Related Categories
General description
- Designed with round bottoms to fit high-speed rotors
- Molding technique produces uniform wall thickness for increased strength
- Uniform diameter (no taper) for better rotor fit and force tolerance
- Lip allows easier retrieval from rotor cavities
- Economical choice - may also be used as test tubes
- Longer-lasting than polycarbonate tubes under conditions of repeated autoclaving
- Autoclavable/Transparent
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)