Skip to Content
Merck
All Photos(1)

Key Documents

T3392

Sigma-Aldrich

Testosterone 3-(O-carboxymethyl)oxime: BSA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
form:
lyophilized powder

Assay

≥98% (TLC)

Quality Level

form

lyophilized powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

extent of labeling

20-30 mol steroid per mol BSA

solubility

phosphate buffer: 1.90-2.10 mg/mL, clear to slightly hazy, colorless (pH 7.50)

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C21H31NO4/c1-20-9-7-14(22-26-12-19(24)25)11-13(20)3-4-15-16-5-6-18(23)21(16,2)10-8-17(15)20/h11,15-18,23H,3-10,12H2,1-2H3,(H,24,25)/t15-,16-,17-,18-,20-,21-/m0/s1

InChI key

VDYLVWGBLQNNAW-ZKHIMWLXSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Guoxin Guo et al.
Molecular and cellular endocrinology, 503, 110671-110671 (2019-12-06)
Although hippocampus-derived androgens play an important role in hippocampal synaptic plasticity, studies at the cellular level have received relatively less attention. Furthermore, the underlying signalling pathways associated with synaptic plasticity remain unclear. Results of the present study demonstrated that testosterone
Yizhou Zhang et al.
Aging, 11(8), 2281-2294 (2019-04-22)
The non-genomic actions of androgen-induced synaptic plasticity have been extensively studied. However, the underlying mechanisms remain controversial. We recently found that testosterone-fetal bovine serum albumin (T-BSA), a cell membrane-impermeable complex, led to a rapid increase in the postsynaptic density 95
Sha Li et al.
Molecular and cellular endocrinology, 414, 82-90 (2015-07-15)
Testosterone (T), the principal androgen, and its metabolite, dihydrotestosterone (DHT), are known to mediate their effects through binding to intracellular androgen receptors (iARs). In addition to their well-known genomic effects, androgens rapidly alter neuronal excitability through a non-genomic pathway mediated

Questions

1–2 of 2 Questions  
  1. What is the molecular weight of Product T3392 Testosterone 3-(O-carboxymethyl)oxime: BSA?

    1 answer
    1. Please see below to calculate the molecular weight of Product T3392, Testosterone 3-(O-carboxymethyl)oxime:BSA.Product T8390 - Testosterone 3-(O-carboxymethyl)oxime  has a formula weight of 361.48 DaltonsProduct T3392 - Testosterone 3-(O-carboxymethyl)oxime: BSA  has 20-30 moles of Testosterone 3-(O-carboxymethyl)oxime per mole of BSA.  The molecular weight for BSA is 66430 Daltons.20 x 361.48 = 7229.630 x 361.48 = 10844.466430 + 7229.6 = 73659.666430 + 10844.4 = 77274.4So the molecular weight range for Product T3392 is 73659.6-77274.4 Daltons.  The lot specific Certificate of Analysis will provide the conjugation ratio.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service