Skip to Content
Merck
All Photos(2)

Documents

T3165

Sigma-Aldrich

7-Methoxy-4-(trifluoromethyl)coumarin

≥99% (TLC)

Synonym(s):

Methyl 4-(trifluoromethyl)umbelliferyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7F3O3
CAS Number:
Molecular Weight:
244.17
Beilstein:
225039
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥99% (TLC)

form

powder

solubility

acetone: 50 mg/mL, clear, colorless to faintly yellow

fluorescence

λex 333 nm; λem 416 nm in methanol

SMILES string

COc1ccc2c(OC(=O)C=C2C(F)(F)F)c1

InChI

1S/C11H7F3O3/c1-16-6-2-3-7-8(11(12,13)14)5-10(15)17-9(7)4-6/h2-5H,1H3

InChI key

HAZHUELNIGDYQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

7-Methoxy-4-(trifluoromethyl)coumarin has been used as a substrate in cytochrome P450 2E1 (CYP2E1) activity assay and aromatase assay.

Biochem/physiol Actions

The enzymatic action of cytochrome P450 2E1 (CYP2E1) on 7-methoxy-4-(trifluoromethyl)coumarin results in a fluorescent product 7-hydroxy-4-(trifluoromethyl)coumarin (HFC) that can be measured at 409/530 nm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebeca Santes-Palacios et al.
Toxicology in vitro : an international journal published in association with BIBRA, 69, 105009-105009 (2020-10-03)
Cytochrome P450 2E1 (CYP2E1) has been proposed as a molecular target in oxidative stress-associated metabolic diseases. Rats are chosen as model organisms in most experiments studying CYP2E1-related toxicity; however, the human relevance of these results remains unclear. To describe differences
Carlo Matera et al.
European journal of pharmacology, 820, 265-273 (2017-12-25)
(±)-3-Methoxy-1-oxa-2,7-diaza-7,10-ethanospiro[4.5]dec-2-ene sesquifumarate (±)-1 was previously characterized as the most selective agonist at α7 neuronal nicotinic acetylcholine receptors in a series of spirocyclic quinuclidinyl-Δ
Vamshi K Manda et al.
Xenobiotica; the fate of foreign compounds in biological systems, 46(2), 117-125 (2015-08-08)
1. Aegle marmelos (bael) is a popular tree in India and other Southeast Asian countries. The fruit is usually consumed as dried, fresh or juice, and is reported to have a high nutritional value and many perceived health benefits. Despite its
Gaku Okamatsu et al.
Xenobiotica; the fate of foreign compounds in biological systems, 47(2), 93-102 (2016-03-10)
1. Little is known about drug metabolism in carnivores. Although the domestic cat (Felis catus) is an obligate carnivore and is the most common companion animal, usage and dosage of many drugs are determined according to information obtained from humans and
Yuka Ono et al.
Xenobiotica; the fate of foreign compounds in biological systems, 49(6), 627-635 (2018-06-01)
1. Compared to information for herbivores and omnivores, knowledge on xenobiotic metabolism in carnivores is limited. The cytochrome P450 2C (CYP2C) subfamily is recognized as one of the most important CYP groups in human and dog. We identified and characterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service