T2532
Tri-O-acetyl-D-galactal
≥90%
Synonym(s):
1,2-Dideoxy-3,4,6-tri-O-acetyl-D-lyxo-1-hexenopyranose
About This Item
Assay
≥90%
mp
34-38 °C (lit.)
SMILES string
CC(=O)OC[C@H]1OC=C[C@@H](OC(C)=O)[C@H]1OC(C)=O
InChI
1S/C12H16O7/c1-7(13)17-6-11-12(19-9(3)15)10(4-5-16-11)18-8(2)14/h4-5,10-12H,6H2,1-3H3/t10-,11-,12-/m1/s1
InChI key
LLPWGHLVUPBSLP-IJLUTSLNSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service