Skip to Content
Merck
All Photos(1)

Key Documents

T1625

Supelco

Testosterone acetate

Synonym(s):

17β-Acetoxy-4-androsten-3-one, 17β-Hydroxy-4-androsten-3-one 17-acetate, 4-Androsten-17β-ol-3-one 17-acetate, Deposteron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
Beilstein:
2062555
EC Number:
MDL number:
UNSPSC Code:
41116107

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])OC(C)=O

Looking for similar products? Visit Product Comparison Guide

General description

Testosterone acetate is an androgen ester and an intermediate in the synthesis of testosterone from progesterone, isolated from microorganisms.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Testosterone and testosterone acetate and the protein and energy metabolism of castrate dogs.
Kochakian DC.
Endocrinology, 21(6), 750-755 (1937)
Metabolic pathways of steroid biosynthesis in ovarian tissue of a teleost, Tilapia aurea.
Eckstein B.
General and Comparative Endocrinology, 14(2), 303-312 (1970)
A Christy Hunter et al.
The Journal of steroid biochemistry and molecular biology, 116(3-5), 171-177 (2009-06-02)
A series of steroids (progesterone, testosterone acetate, 17beta-acetoxy-5 alpha-androstan-3-one, testosterone and androst-4-en-3,17-dione) have been incubated with the thermophilic ascomycete Myceliophthora thermophila CBS 117.65. A wide range of biocatalytic activity was observed with modification at all four rings of the steroid
S B Addo et al.
Steroids, 54(3), 257-269 (1989-09-01)
Non-polar extracts of sera from human males contain immunoreactive testosterone in a form that is released by mild alkaline hydrolysis. The non-polar derivative shows no immunoreactivity with testosterone antibody prior to hydrolysis. Hydrolyzed non-polar serum extracts from ten adult male
Mei Hua Sui et al.
Shi yan sheng wu xue bao, 36(1), 43-48 (2003-05-20)
In order to study the influence of pregnancy and hormone on the lymphatic stomata of guinea pig's ovary bursa, the lymphatic stomata of ovary bursa were observed during pregnancy and by using exogenous hormone, including serum gonadotrophin (PMSG) combined with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service