Skip to Content
Merck
All Photos(1)

Key Documents

T1516

Sigma-Aldrich

DL-Trihexyphenidyl hydrochloride

solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31NO · HCl
CAS Number:
Molecular Weight:
337.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

methanol: soluble 50 mg/ml, clear to hazy, colorless
H2O: soluble

SMILES string

Cl[H].OC(CCN1CCCCC1)(C2CCCCC2)c3ccccc3

InChI

1S/C20H31NO.ClH/c22-20(18-10-4-1-5-11-18,19-12-6-2-7-13-19)14-17-21-15-8-3-9-16-21;/h1,4-5,10-11,19,22H,2-3,6-9,12-17H2;1H

InChI key

QDWJJTJNXAKQKD-UHFFFAOYSA-N

Application

DL-Trihexyphenidyl hydrochloride has been used to develop a capillary zone electrophoretic method with end-column electrochemiluminescence (ECL)-based detection technique.

Biochem/physiol Actions

DL-trihexyphenidyl hydrochloride (Benzhexol hydrochloride) is known to function as an antimuscarinic agent and a centrally acting anticholinergic. It can be used for controlling the symptoms of all Parkinsonism types, such as the idiopathic, and postencephalitic arteriosclerotic forms. Benzhexol hydrochloride exerts inhibitory functions on the parasympathetic nervous system.
Muscarinic acetylcholine receptor antagonist

Preparation Note

DL-Trihexyphenidyl hydrochloride is soluble in water. It dissolves in methanol at 50 mg/ml to yield a clear to hazy, colorless solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Hayakawa et al.
Clinical and experimental pharmacology & physiology, 26(5-6), 421-425 (1999-07-01)
1. Effects of anti-Parkinsonian drugs on neurobehavioural changes induced by bilateral lesions of dopaminergic neurons were investigated in rats. 2. Dopaminergic neurons in rats were lesioned bilaterally by injection of 6-hydroxydopamine (6-OHDA; 8 micrograms) into the medial forebrain bundle at
Determination of benzhexol hydrochloride by capillary zone electrophoresis with an end-column electrochemiluminescence detection.
Yan, J., et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 76(1), 11-16 (2004)
H E Shannon et al.
The Journal of pharmacology and experimental therapeutics, 290(2), 901-907 (1999-07-20)
The purpose of our studies was to determine the effects of muscarinic receptor agonists on conditioned avoidance responding in the rat. Rats were trained to avoid or escape an electric shock delivered to the feet in a discrete trial procedure.
Y Kimura et al.
Brain research, 834(1-2), 6-12 (1999-07-17)
An amnesic effect of anticholinergic drugs was previously described from several behavioral studies. We examined this effect induced by trihexyphenidyl and biperiden, clinically used in the parkinsonism and schizophrenic patients, by using passive avoidance tasks. Both of these drugs (0.1-10

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service