T1501
DL-Thyronine
powder
Synonym(s):
3-(p-[p-Hydroxyphenoxy]phenyl)-DL-alanine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H15NO4
CAS Number:
Molecular Weight:
273.28
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
form
powder
color
off-white to gray
SMILES string
NC(Cc1ccc(Oc2ccc(O)cc2)cc1)C(O)=O
InChI
1S/C15H15NO4/c16-14(15(18)19)9-10-1-5-12(6-2-10)20-13-7-3-11(17)4-8-13/h1-8,14,17H,9,16H2,(H,18,19)
InChI key
KKCIOUWDFWQUBT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Huai Yang et al.
Cell and tissue research, 374(2), 303-315 (2018-08-02)
Tissue-engineered urethra with autologous cells seeded on biodegradable scaffolds offers an alternative for lower urinary tract reconstruction. Rabbit is most commonly used as an animal model in urethra and bladder tissue repair. The goal of this study is to characterize
Min Kyong Moon et al.
Molecular and cellular endocrinology, 436, 50-58 (2016-07-28)
Thyroid-stimulating hormone (TSH) receptor is expressed in extrathyroidal tissues such as hepatocytes, adipocytes, and skeletal muscle, which suggests a possible novel role of TSH in various metabolic processes in extrathyroidal tissues independent of thyroid hormones. We investigated whether TSH has
Ailin Lepletier et al.
Cell reports, 27(13), 3887-3901 (2019-06-27)
A key feature of immune functional impairment with age is the progressive involution of thymic tissue responsible for naive T cell production. In this study, we identify two major phases of thymic epithelial cell (TEC) loss during aging: a block in
Lara Jobeili et al.
Aging, 9(11), 2302-2315 (2017-11-28)
Skin is constantly exposed to environmental factors such as pollutants, chemicals and ultra violet radiation (UV), which can induce premature skin aging and increase the risk of skin cancer. One strategy to reduce the effect of oxidative stress produced by
Yusuf B Johari et al.
Biotechnology and bioengineering, 118(5), 2001-2015 (2021-02-14)
Age-related macular degeneration (AMD) associated with dysfunction of retinal pigment epithelial (RPE) cells is the most common cause of untreatable blindness. To advance gene therapy as a viable treatment for AMD there is a need for technologies that enable controlled
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service