Skip to Content
Merck
All Photos(1)

Key Documents

T1129

Sigma-Aldrich

DL-Tryptophan ethyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O2 · HCl
CAS Number:
Molecular Weight:
268.74
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

Cl.[H]n1cc(CC(N)C(=O)OCC)c2ccccc12

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Fukushima et al.
Urological research, 10(3), 115-118 (1982-01-01)
The effect of partial cystectomy on the occurrence of pre-neoplastic lesions, papillary or nodular hyperplasia (PN hyperplasia), of the bladder in male F344 rats was studied in an experiment in which bladder carcinogens and promoters were given to the rats
F Kilár et al.
Electrophoresis, 16(8), 1510-1518 (1995-08-01)
Enantiomers can be separated by using human serum transferrin as a chiral phase. With the help of the native protein we were able to separate enantiomers with high efficiency, using a low ionic strength 2-(N-morpholino)ethanesulfonic acid (MES) buffer, pH 6
R P Kwok et al.
Neuroendocrinology, 45(4), 267-273 (1987-04-01)
In this study we investigated the effect of insulin on striatal 5-hydroxytryptamine and tryptamine in streptozotocin-diabetic and in normal rats. Streptozotocin-diabetic rats show a reduction in rat striatal tryptophan, 5-hydroxytryptamine, and 5-hydroxyindole acetic acid, an effect observed at 7 or
A J Jonas et al.
The Journal of clinical investigation, 84(1), 200-204 (1989-07-01)
Tryptophan ethyl ester, a lipid-soluble tryptophan derivative, was used to bypass defective gastrointestinal neutral amino acid transport in a child with Hartnup disease. The child's baseline tryptophan concentrations in serum (20 +/- 6 microM) and cerebrospinal fluid (1.0 +/- 0.2
Jian Xiang Zhang et al.
Journal of controlled release : official journal of the Controlled Release Society, 116(3), 322-329 (2006-11-18)
The effects of copolymer composition, drug structure and initial drug feed on drug loading of polymeric micelles based on amphiphilic polyphosphazenes were investigated. It was found that the drug loading capacity of micelles based on this type of amphiphilic copolymers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service