Skip to Content
Merck
All Photos(1)

Documents

T0652

SAFC

Tunicamycin A2 homolog

~90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H60N4O16
CAS Number:
Molecular Weight:
816.89
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

~90% (HPLC)

storage temp.

−20°C

InChI

1S/C37H60N4O16/c1-3-4-5-6-7-8-9-10-11-12-13-14-23(45)39-26-30(50)27(47)21(54-36(26)57-35-25(38-19(2)43)29(49)28(48)22(18-42)55-35)17-20(44)33-31(51)32(52)34(56-33)41-16-15-24(46)40-37(41)53/h13-16,20-22,25-36,42,44,47-52H,3-12,17-18H2,1-2H3,(H,38,43)(H,39,45)(H,40,46,53)/b14-13+

InChI key

MMDMXWRRAOLMDC-BUHFOSPRSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service