Key Documents
T0566
Dounce tissue grinder tube
working volume 7 mL
Select a Size
1 EA
$126.00
$126.00
Available to ship on07 April 2025Details
Select a Size
Change View
1 EA
$126.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
$126.00
Available to ship on07 April 2025Details
material
glass
packaging
pack of 1 ea
working volume
7 mL
General description
Designed primarily for cellular work where the nucleus remains intact after homogenization.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. I would like to ask you regarding the product https://www.sigmaaldrich.com/GR/en/product/sigma/t0566 Which pestle is suitable for the particular grinder?
    1 answer
    1. There are two pestle options for the 7 mL tissue grinder.
      See below for a link to each product.
      P1235, Small Clearance Pestle
      https://www.sigmaaldrich.com/product/sigma/p1235
      P0610, Large Clearance Pestle
      https://www.sigmaaldrich.com/product/sigma/p0610
      Helpful?