Skip to Content
Merck
All Photos(1)

Key Documents

T0453

Sigma-Aldrich

TAOK1 (1-314), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FLJ14314, KIAA1361, MAP3K16, MARKK, PSK2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$537.00

$537.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$537.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

$537.00


Estimated to ship on18 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1420-1921 nmol/min·mg

mol wt

~63 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TAOK1(57551)

Biochem/physiol Actions

TAOK1 is a serine/threonine-protein kinase involved in regulation of the p38-containing stress-responsive MAP kinase pathway and extracellular signal-regulated protein kinase (ERK) kinases (MEKs). The activation of and binding to MEK3 by TAOK1 implicates TAOK1 in the regulation of the p38-containing stress-responsive MAP kinase pathway. A microtubule affinity-regulating kinase kinase, TAOK1 (also known as MARKK) is an important regulator of mitotic progression, required for both chromosome congression and checkpoint-induced anaphase delay.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viji M Draviam et al.
Nature cell biology, 9(5), 556-564 (2007-04-10)
Defects in chromosome-microtubule attachment trigger spindle-checkpoint activation and delay mitotic progression. How microtubule attachment is sensed and integrated into the steps of checkpoint-signal amplification is poorly understood. In a functional genomic screen targeting human kinases and phosphatases, we identified a
M Hutchison et al.
The Journal of biological chemistry, 273(44), 28625-28632 (1998-10-24)
Several components of the budding yeast pheromone-response pathway are conserved in mammalian mitogen-activated protein (MAP) kinase pathways. Thus, we used degenerate oligonucleotides derived from the sequence of the Saccharomyces cerevisiae protein kinase Ste20p to amplify related sequences from the rat.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service